Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgbd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_171824.2 | NP_741958.1 | 7 | 1217 | ||
GenBank mRNA | BC094384.1 | - | 1215 | 30606749 | ||
BM941957.1 | - | 1 | 622 |