Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35f3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175434.3 | NP_780643.2 | 2 | 528 | ||
GenBank mRNA | BC115859.1 | - | 2 | 374 | 40057292 | |
BC115965.1 | - | 2 | 374 | 40057290 |