Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D130043K22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK051381.1 | - | 17, 18 | 3005 | ||
AK122246.1 | - | 18, 19 | 3141 | |||
AK148668.1 | - | 16, 17 | 2748 | |||
BC115723.1 | - | 18, 19 | 3060 | 40086618 | ||
BC115940.1 | - | 18, 19 | 3060 | 40086616 |