Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp526 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175436.4 | NP_780645.1 | 3 | 499 | ||
GenBank mRNA | AK050839.1 | - | 3 | 500 | ||
AK051798.1 | - | 3 | 496 | |||
AK173308.1 | - | 476 | ||||
BC052548.1 | - | 2 | 453 |