Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pank3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145962.2 | NP_666074.1 | 2 | 665 | ||
GenBank mRNA | AK039865.1 | - | 2 | 667 | ||
AK136305.1 | - | 2 | 355 | |||
AK163377.1 | - | 1 | 52 | |||
BC027089.1 | - | 2 | 277 | 5355443 | ||
BC032188.1 | - | 2 | 711 |