Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004364.2 | NP_001004364.2 | 3 | 573 | ||
NM_001098168.1 | NP_001091637.1 | 3 | 573 | |||
NM_001135192.1 | NP_001128664.1 | 3 | 573 | |||
GenBank mRNA | AK018309.1 | - | 1433 | |||
AK147622.1 | - | 3 | 581 | |||
AK147656.1 | - | 3 | 519 | |||
AK162242.1 | - | 2239 | ||||
AK172944.1 | - | 3 | 385 | |||
BC080847.1 | - | 3 | 573 | 6852788 | ||
BC096022.1 | - | 3 | 341 | 30542525 | ||
CD351639.1 | - | 3 | 573 |