Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164559.1 | NP_001158031.1 | 5 | 658 | ||
NM_001164560.1 | NP_001158032.1 | 5 | 582 | |||
NM_198020.2 | NP_932137.2 | 5 | 685 | |||
GenBank mRNA | AK032945.1 | - | 4 | 831 | ||
AK078395.1 | - | 4 | 861 | |||
AK084775.1 | - | 5 | 623 | |||
AK139463.1 | - | 5 | 617 | |||
AK155251.1 | - | 5 | 650 | |||
AK159467.1 | - | 5 | 623 | |||
AK170412.1 | - | 5 | 547 | |||
AK171002.1 | - | 5 | 650 | |||
BC027404.1 | - | 5 | 654 | 4950980 |