Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prpsap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164242.1 | NP_001157714.1 | 10 | 1101 | ||
NM_001164243.1 | NP_001157715.1 | 10 | 1144 | |||
NM_001164244.1 | NP_001157716.1 | 9 | 1048 | |||
NM_144806.2 | NP_659055.1 | 11 | 1197 | |||
GenBank mRNA | AK038680.1 | - | 11 | 1163 | ||
AK076811.1 | - | 11 | 1151 | |||
AK077330.1 | - | 11 | 1150 | |||
BC023178.1 | - | 11 | 1182 | 4947226 | ||
BC051438.1 | - | 10 | 1068 | 6311509 | ||
BC106185.1 | - | 10 | 1030 | 6512229 |