Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prpsap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164242.1 | NP_001157714.1 | 6 | 622 | ||
NM_001164243.1 | NP_001157715.1 | 6 | 665 | |||
NM_001164244.1 | NP_001157716.1 | 5 | 569 | |||
NM_144806.2 | NP_659055.1 | 7 | 718 | |||
GenBank mRNA | AK038680.1 | - | 7 | 684 | ||
AK076811.1 | - | 7 | 672 | |||
AK077330.1 | - | 7 | 671 | |||
BC023178.1 | - | 7 | 703 | 4947226 | ||
BC051438.1 | - | 6 | 589 | 6311509 | ||
BC106185.1 | - | 6 | 551 | 6512229 |