Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153391.2 | NP_700440.2 | 26 | 2983 | ||
GenBank mRNA | AK052303.1 | - | 26 | 2963 | ||
AK129413.1 | - | 18 | 2072 | |||
AK147970.1 | - | 26 | 2983 | |||
AY029258.1 | - | 26 | 3053 | |||
BC046432.1 | - | 11 | 1185 | |||
BC060134.1 | - | 26 | 2977 | |||
BC082301.1 | - | 25 | 2863 |