Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pddc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172116.4 | NP_742114.3 | 5 | 457 | ||
GenBank mRNA | AK081187.1 | - | 5 | 417 | ||
AK084273.1 | - | 5 | 413 | |||
AK088159.1 | - | 5 | 433 | |||
AK140975.1 | - | 5 | 408 | |||
AK150323.1 | - | 3 | 1127 | |||
AK150354.1 | - | 3 | 1127 | |||
AK151259.1 | - | 3 | 1127 | |||
AK151794.1 | - | 3 | 1127 | |||
AK151959.1 | - | 3 | 1127 | |||
AK153110.1 | - | 3 | 1127 | |||
AK161205.1 | - | 4 | 1260 | |||
AK181635.1 | - | 2 | 100 | |||
BC023835.1 | - | 5 | 427 | 5345186 |