Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lgi3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145219.4 | NP_660254.1 | 8 | 1326 | ||
GenBank mRNA | AF515590.1 | - | 8 | 1308 | ||
AK047495.1 | - | 8 | 1278 | |||
AK049831.1 | - | 8 | 1278 | |||
AK087125.1 | - | 9 | 1465 | |||
AK132296.1 | - | 8 | 1320 | |||
AY174077.1 | - | 8 | 1308 | |||
BC055315.1 | - | 8 | 1186 | 5686778 | ||
BC061460.1 | - | 8 | 1186 | 5686778 |