Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tagln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178598.2 | NP_848713.1 | 4 | 501 | ||
GenBank mRNA | AF149291.1 | - | 4 | 492 | ||
AF465519.1 | - | 4 | 438 | |||
AK151576.1 | - | 4 | 501 | |||
AK167395.1 | - | 4 | 486 | |||
AK167922.1 | - | 4 | 483 | |||
AK171088.1 | - | 4 | 469 | |||
AK172892.1 | - | 4 | 440 | |||
AK195747.1 | - | 3 | 264 | |||
BC009076.1 | - | 4 | 483 | 2654381 | ||
BC049861.1 | - | 4 | 700 | 4981433 |