Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK145234.1 | - | 6 | 1162 | ||
AK169645.1 | - | 3 | 410 | |||
BC005578.1 | - | 6 | 1102 | 3968225 | ||
BC051257.1 | - | 6 | 1115 | 6530488 | ||
BC057668.1 | - | 6 | 1489 | |||
CT010387.1 | - | 5 | 1049 | |||
KJ650229.1 | - | 5 | 1052 | |||
U60087.1 | - | 5 | 1052 | |||
U60088.1 | - | 5 | 1052 | |||
U60089.1 | - | 5 | 1052 | |||
U60090.1 | - | 5 | 1052 | |||
U60091.1 | - | 5 | 1052 | |||
U60092.1 | - | 5 | 1049 |