Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tapbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025313.1 | NP_001020484.1 | 5 | 1367 | ||
NM_009318.2 | NP_033344.1 | 5 | 1367 | |||
GenBank mRNA | AF043943.1 | - | 5 | 1225 | ||
AF106278.1 | - | 5 | 1157 | |||
AJ316613.2 | - | 5 | 1157 | |||
AK014566.1 | - | 5 | 1222 | |||
AK054174.1 | - | 2 | 542 | |||
AK077299.1 | - | 5 | 1191 | |||
AK151134.1 | - | 5 | 1188 | |||
AK151807.1 | - | 5 | 1170 | |||
AK151878.1 | - | 5 | 1184 | |||
AK152019.1 | - | 5 | 1183 | |||
AK152726.1 | - | 2 | 636 | |||
AK169697.1 | - | 5 | 1188 | |||
AK170528.1 | - | 5 | 1178 | |||
AY734468.1 | - | 5 | 1141 | |||
AY734469.1 | - | 5 | 1140 | |||
AY734471.1 | - | 5 | 1155 | |||
AY734475.1 | - | 4 | 1123 | |||
AY734476.1 | - | 1 | 108 | |||
BC013236.1 | - | 4 | 965 | |||
BC015074.1 | - | 5 | 1180 | 4223922 | ||
KJ650208.1 | - | 5 | 1157 |