Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tarbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253795.1 | NP_001240724.1 | 5 | 752 | ||
NM_009319.3 | NP_033345.2 | 7 | 1025 | |||
GenBank mRNA | AK143188.1 | - | 6 | 924 | ||
AK144205.1 | - | 5 | 590 | |||
BC002028.1 | - | 7 | 738 | 3489376 | ||
BC053432.1 | - | 2 | 1525 | |||
U79962.1 | - | 7 | 758 |