Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011535.3 | NP_035665.2 | 7 | 2440 | ||
NM_198052.2 | NP_932169.1 | 7 | 2576 | |||
GenBank mRNA | AK052453.1 | - | 7 | 2655 | ||
AK087128.1 | - | 1 | 208 | |||
AK145436.1 | - | 7 | 2357 | |||
AK157247.1 | - | 6 | 2141 | |||
AK172118.1 | - | 6 | 2144 | |||
BC096551.1 | - | 7 | 2576 | 30547736 |