Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sox30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173384.2 | NP_775560.1 | 5 | 1990 | ||
GenBank mRNA | AY005801.1 | - | 5 | 1990 |