Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tet2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040400.2 | NP_001035490.2 | 3333 | |||
GenBank mRNA | AK151359.1 | - | 1 | 323 | ||
AK152297.1 | - | 1 | 323 | |||
AK153251.1 | - | 1 | 323 | |||
AK153311.1 | - | 1 | 323 | |||
AK153460.1 | - | 1 | 323 | |||
DQ079067.1 | - | 3 | 3311 | |||
GU079949.1 | - | 1 | 2957 |