Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172525.2 | NP_766113.1 | 23 | 3217 | ||
GenBank mRNA | AK132716.1 | - | 18 | 2603 | ||
BC031572.1 | - | 12 | 1884 | |||
BC040387.1 | - | 23 | 3241 | 3599962 |