Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pstk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039534.1 | NP_001034623.1 | 3 | 713 | ||
GenBank mRNA | AI173292.1 | - | 1 | 209 | ||
AK077585.1 | - | 3 | 714 | |||
BC030313.1 | - | 1 | 1205 | |||
BC110381.1 | - | 3 | 689 | 5149569 | ||
BC115503.2 | - | 3 | 714 |