Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Papd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164497.1 | NP_001157969.1 | 10, 11 | 1653 | ||
NM_001164498.1 | NP_001157970.1 | 10, 11 | 1627 | |||
NM_001164499.1 | NP_001157971.1 | 9, 10 | 1524 | |||
GenBank mRNA | AK077553.1 | - | 10, 11 | 1655 | ||
BC059053.1 | - | 10, 11 | 1625 | |||
BC080314.1 | - | 9, 10 | 1228 | |||
BC144796.1 | - | 10, 11 | 1611 | 9053344 | ||
BC144797.1 | - | 9, 10 | 1482 | 9053345 | ||
BC145737.1 | - | 10, 11 | 1611 | 40131040 |