Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vgll2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300957.1 | NP_001287886.1 | 1, 2 | 307 | ||
NM_153786.2 | NP_722481.1 | 1, 2 | 307 | |||
GenBank mRNA | AF542181.1 | - | 1, 2 | 123 | ||
AJ578054.1 | - | 1, 2 | 125 | |||
AK048390.1 | - | 1, 2 | 279 | |||
AK052987.1 | - | 1, 2 | 282 | |||
BC133696.1 | - | 1, 2 | 123 | 40081288 |