Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il1f10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153077.2 | NP_694717.1 | 4 | 443 | ||
GenBank mRNA | AY071844.1 | - | 4 | 443 | ||
BC107311.1 | - | 4 | 435 | 40053322 | ||
BC107312.1 | - | 4 | 439 | 40053325 |