Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prr14l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194340.2 | NP_919321.2 | 10, 11 | 5911 | ||
GenBank mRNA | AK049285.1 | - | 3, 4 | 243 | ||
AY372183.1 | - | 5, 6 | 1132 | |||
BC059908.1 | - | 3, 4 | 343 | |||
BC094374.1 | - | 5, 6 | 1458 | 30531815 |