Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A3galt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009819.2 | NP_001009819.1 | 2 | 796 | ||
GenBank mRNA | AK132334.1 | - | 2 | 797 | ||
AY491982.1 | - | 2 | 145 |