Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc28a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144820.3 | NP_659069.2 | 2 | 213 | ||
GenBank mRNA | AK028075.1 | - | 2 | 182 | ||
AK148447.1 | - | 2 | 214 | |||
BC013553.1 | - | 2 | 210 | |||
BC027759.1 | - | 2 | 221 | 5346317 |