Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc28a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144820.3 | NP_659069.2 | 4 | 549 | ||
GenBank mRNA | AK028075.1 | - | 4 | 518 | ||
AK046998.1 | - | 2 | 173 | |||
AK148447.1 | - | 4 | 550 | |||
BC013553.1 | - | 4 | 547 | |||
BC027759.1 | - | 4 | 557 | 5346317 |