Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AI317395 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144821.4 | NP_659070.2 | 4 | 1607 | ||
GenBank mRNA | AK089961.1 | - | 4 | 1610 | ||
BC018406.1 | - | 4 | 1511 | 4196864 |