Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D930020B18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177335.4 | NP_796309.3 | 4, 5 | 1000 | ||
GenBank mRNA | AK033441.1 | - | 5, 6 | 1177 |