Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfap36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025740.3 | NP_080016.1 | 2 | 303 | ||
GenBank mRNA | AK010468.1 | - | 2 | 330 | ||
AK016479.1 | - | 2 | 339 | |||
AK075858.1 | - | 2 | 304 | |||
AK219590.1 | - | 2 | 93 | |||
BC005731.1 | - | 2 | 273 | 3595965 |