Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033597.1 | NP_001028769.1 | 19 | 1960 | ||
NM_001033598.1 | NP_001028770.1 | 19 | 1929 | |||
NM_001033599.1 | NP_001028771.1 | 19 | 2135 | |||
NM_144823.4 | NP_659072.3 | 19 | 1960 | |||
GenBank mRNA | AB073974.1 | - | 18 | 1804 | ||
AK122384.1 | - | 8 | 3726 | |||
AK147283.1 | - | 19 | 1930 | |||
AK147421.1 | - | 19 | 1961 | |||
AY167035.1 | - | 19 | 1930 | |||
AY786360.1 | - | 21 | 2478 | |||
AY786361.1 | - | 19 | 1910 | |||
AY786362.1 | - | 19 | 2102 | |||
AY786363.1 | - | 19 | 1955 | |||
BC016114.1 | - | 19 | 1886 | 4481949 | ||
BC022959.1 | - | 19 | 1960 | 5362660 |