Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acsl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033597.1 | NP_001028769.1 | 5 | 598 | ||
NM_001033598.1 | NP_001028770.1 | 5 | 567 | |||
NM_001033599.1 | NP_001028771.1 | 5 | 773 | |||
NM_144823.4 | NP_659072.3 | 5 | 598 | |||
GenBank mRNA | AB073974.1 | - | 4 | 442 | ||
AK032479.1 | - | 6 | 857 | |||
AK039380.1 | - | 5 | 684 | |||
AK147283.1 | - | 5 | 568 | |||
AK147421.1 | - | 5 | 599 | |||
AY167035.1 | - | 5 | 568 | |||
AY786360.1 | - | 7 | 1116 | |||
AY786361.1 | - | 5 | 548 | |||
AY786362.1 | - | 5 | 740 | |||
AY786363.1 | - | 5 | 593 | |||
BC016114.1 | - | 5 | 524 | 4481949 | ||
BC022959.1 | - | 5 | 598 | 5362660 |