Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045521.1 | NP_001038986.1 | 3, 4 | 605 | ||
GenBank mRNA | AK145810.1 | - | 3, 4 | 610 | ||
BC147400.1 | - | 3, 4 | 601 | 9056455 | ||
BC147401.1 | - | 3, 4 | 601 | 9056447 | ||
BY736706.1 | - | 3, 4 | 605 |