Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmgt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175002.2 | NP_778167.1 | 2 | 701 | ||
GenBank mRNA | AK004099.1 | - | 2 | 538 | ||
AK131977.1 | - | 2 | 591 | |||
BC025076.1 | - | 1 | 472 | 4459181 | ||
BC096408.1 | - | 3 | 316 | |||
BC096583.1 | - | 2 | 645 | 5025927 | ||
BI134823.1 | - | 1 | 170 | |||
EU069462.1 | - | 1 | 359 |