Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099288.1 | NP_001092758.1 | 6, 7 | 795 | ||
NM_175003.3 | NP_778168.2 | 6, 7 | 795 | |||
GenBank mRNA | AK053220.1 | - | 6, 7 | 523 | ||
AK079081.1 | - | 1, 2 | 96 | |||
BC056366.1 | - | 6, 7 | 795 | 6825221 | ||
BC059911.1 | - | 6, 7 | 795 | 6825221 |