Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291049.1 | NP_001277978.1 | 12 | 1860 | ||
NM_173754.4 | NP_776115.2 | 12 | 1860 | |||
GenBank mRNA | AK042217.1 | - | 12 | 1861 | ||
AK083271.1 | - | 12 | 1698 | |||
BC021474.1 | - | 11 | 1150 | 5343429 |