Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cntrob | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172560.3 | NP_766148.2 | 17 | 3344 | ||
GenBank mRNA | AK034947.1 | - | 3 | 968 | ||
AK036710.1 | - | 16 | 3164 | |||
AK147774.1 | - | 17 | 3234 | |||
BC058512.1 | - | 17 | 2177 | 6817014 |