Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlgn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198862.2 | NP_942562.2 | 5 | 1425 | ||
GenBank mRNA | AK136883.1 | - | 6 | 1991 | ||
AK173159.1 | - | 5 | 1425 | |||
BE952199.1 | - | 5 | 629 |