Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gltpd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146020.1 | NP_666132.1 | 1 | 51 | ||
GenBank mRNA | AK050107.1 | - | 1 | 65 | ||
AK050212.1 | - | 1 | 65 | |||
BC025515.1 | - | 1 | 50 | 5309987 | ||
BC030735.1 | - | 1 | 51 | 5126172 |