Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camta2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190376.1 | NP_001177305.1 | 20, 21 | 3567 | ||
NM_178116.4 | NP_835217.1 | 20, 21 | 3582 | |||
GenBank mRNA | AK122400.1 | - | 18, 19 | 3223 | ||
BC027039.1 | - | 9, 10 | 1291 | |||
BC049133.1 | - | 20, 21 | 3411 | 6827800 | ||
BC056395.1 | - | 20, 21 | 3396 | 6852349 |