Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BC030499 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287206.1 | NP_001274135.1 | 8, 9 | 750 | ||
GenBank mRNA | BC037104.1 | - | 7, 8 | 1077 | ||
BC059191.1 | - | 7, 8 | 1088 |