Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tada2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172562.3 | NP_766150.1 | 8 | 663 | ||
GenBank mRNA | AK142262.1 | - | 8 | 695 | ||
AK157938.1 | - | 8 | 656 | |||
BC005610.1 | - | 5 | 390 | |||
BC025448.1 | - | 3 | 155 | |||
BC038821.1 | - | 8 | 666 | 3990023 |