Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cisd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085500.2 | NP_001078969.2 | 2 | 331 | ||
GenBank mRNA | AA987107.1 | - | 2 | 346 | ||
BC056217.1 | - | 2 | 326 | |||
BC147739.1 | - | 3 | 426 | 9056688 | ||
BC147742.1 | - | 3 | 426 | 9056691 | ||
BC157903.1 | - | 3 | 369 |