Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl5c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207231.1 | NP_997114.1 | 3 | 540 | ||
GenBank mRNA | AK032519.1 | - | 3 | 297 | ||
AK154137.1 | - | 3 | 260 | |||
AK171194.1 | - | 3 | 551 | |||
BC065791.1 | - | 3 | 540 | 3824354 |