Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhh3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146030.2 | NP_666142.1 | 9 | 1718 | ||
GenBank mRNA | AK154510.1 | - | 9 | 1734 | ||
AK164128.1 | - | 9 | 1737 | |||
BC020025.1 | - | 9 | 1718 | 3490992 | ||
BC024538.1 | - | 2 | 155 |