Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pyy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145435.1 | NP_663410.1 | 2 | 72 | ||
GenBank mRNA | AK050518.1 | - | 2 | 129 | ||
BC010821.1 | - | 2 | 72 | 4218355 |