Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd300a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170758.3 | NP_739564.1 | 5, 6 | 1002 | ||
GenBank mRNA | AB065156.1 | - | 5, 6 | 721 | ||
AB091765.1 | - | 5, 6 | 728 | |||
AB091766.1 | - | 5, 6 | 724 | |||
AB095675.1 | - | 5, 6 | 825 | |||
AK045869.1 | - | 5, 6 | 1002 | |||
AK053160.1 | - | 5, 6 | 678 | |||
AY457054.1 | - | 5, 6 | 678 | |||
BC139296.1 | - | 5, 6 | 938 | 8862318 | ||
BC139297.1 | - | 5, 6 | 938 | 8862319 |