Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab40b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139147.3 | NP_631886.2 | 1, 2 | 258 | ||
GenBank mRNA | AB232638.1 | - | 1, 2 | 146 | ||
AF425643.1 | - | 1, 2 | 146 | |||
BC096601.1 | - | 1, 2 | 252 | 4974190 | ||
BY138215.1 | - | 1, 2 | 199 | |||
BY261509.1 | - | 1, 2 | 258 |