Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Terf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286628.1 | NP_001273557.1 | 3 | 465 | ||
NM_009352.3 | NP_033378.1 | 3 | 465 | |||
GenBank mRNA | AB109563.1 | - | 3 | 445 | ||
AK021235.1 | - | 3 | 455 | |||
AK036615.1 | - | 3 | 468 | |||
AK132022.1 | - | 3 | 448 | |||
BB652245.1 | - | 3 | 466 | |||
BC034866.1 | - | 3 | 453 | 3470277 | ||
U65586.1 | - | 3 | 445 |